Assay Details
| Gene Symbol: Gcsam | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159297.1 | NP_001152769.1 | 4 - 5 | 297 | 111 | |
| NM_008099.4 | NP_032125.3 | 3 - 4 | 231 | 111 | ||
| GenBank mRNA | AK036553.1 | - | 3 - 4 | 249 | 111 | |
| AY500830.1 | - | 4 - 5 | 205 | 111 | ||
| BC099585.1 | - | 3 - 4 | 195 | 30289367 | 111 | |
| U13263.1 | - | 3 - 4 | 232 | 111 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gcsam
Genomic Map