Assay Details
| Gene Symbol: Mgrn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252437.1 | NP_001239366.1 | 14 - 15 | 1750 | 79 | |
| NM_029657.4 | NP_083933.1 | 14 - 15 | 1747 | 79 | ||
| GenBank mRNA | AK011747.2 | - | 14 - 15 | 1540 | 79 | |
| AK034100.1 | - | 5 - 6 | 578 | 79 | ||
| AK088533.1 | - | 14 - 15 | 1530 | 79 | ||
| AK129161.1 | - | 11 - 12 | 1158 | 79 | ||
| AK150176.1 | - | 15 - 16 | 1583 | 79 | ||
| AK153407.1 | - | 14 - 15 | 1571 | 79 | ||
| AK203289.1 | - | 2 - 3 | 130 | 79 | ||
| BC046830.1 | - | 14 - 15 | 1596 | 6811606 | 79 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mgrn1
Genomic Map