| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1600000328.mm.1 |
NM_001252437.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600000328.mm.1 |
NM_001252437.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600000328.mm.1 |
NM_029657.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600000328.mm.1 |
NM_029657.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600000328.mm.1 |
TC1600000038.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1600000331.mm.1 |
NM_001252437.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600000331.mm.1 |
NM_001252437.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600000331.mm.1 |
NM_029657.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600000331.mm.1 |
NM_029657.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600000331.mm.1 |
TC1600000038.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1600000329.mm.1 |
NM_001252437.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600000329.mm.1 |
NM_001252437.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600000329.mm.1 |
NM_029657.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600000329.mm.1 |
NM_029657.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600000329.mm.1 |
TC1600000038.mm.1 |
MTA TC |