Assay Details
| Gene Symbol: Mgrn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252437.1 | NP_001239366.1 | 5 - 6 | 849 | 80 | |
| NM_029657.4 | NP_083933.1 | 5 - 6 | 846 | 80 | ||
| GenBank mRNA | AK011747.2 | - | 5 - 6 | 639 | 80 | |
| AK088533.1 | - | 5 - 6 | 629 | 80 | ||
| AK129161.1 | - | 2 - 3 | 257 | 80 | ||
| AK150176.1 | - | 6 - 7 | 682 | 80 | ||
| AK153407.1 | - | 5 - 6 | 670 | 80 | ||
| BC046830.1 | - | 5 - 6 | 695 | 6811606 | 80 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mgrn1
Genomic Map