Assay Details
| Gene Symbol: Vgll3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028572.1 | NP_082848.1 | 2 - 3 | 408 | 82 | |
| GenBank mRNA | BC042696.1 | - | 2 - 3 | 333 | 82 | |
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_65_P04847 | AK031197.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_65_P04847 | NM_028572.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 297358 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 435325 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 808831 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 454913 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 249059 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 316670 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 138374 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 185834 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 847274 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 668040 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 702819 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 714653 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 882550 | AK156006.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 645094 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 404579 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 10161 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 381162 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 49889 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 210192 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 862785 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 644511 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 29779 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 870129 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 725233 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 535661 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 62765 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 919541 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 77864 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 103623 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 74215 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 151087 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 44558 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_65_P04847 | AK031197.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_65_P04847 | NM_028572.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08627246 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08627169 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08627263 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08627214 | - | - |
| Whole Mouse Genome G4122A | A_51_P309234 | BC042696.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P309234 | AK007165.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P309234 | AK031197.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P309234 | NM_028572.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P08627214 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P08627263 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30826633 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30826654 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30826549 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12619301 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30826616 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30826641 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30826797 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30826605 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30826611 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12596199 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30826615 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30826828 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30826693 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30826820 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30826695 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30826623 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30826656 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12501170 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12596202 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12922052 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12500669 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30826542 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30826592 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30826800 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30826674 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30826697 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30826755 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30826761 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30826796 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12699037 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30826664 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30826634 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30826628 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30826583 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30826668 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30826690 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30826679 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30826551 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30826574 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30826562 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30826557 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30826817 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30826618 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12889323 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30826849 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30826558 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30826759 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30826658 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30826823 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30826824 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30826591 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30826804 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30826802 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30826832 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30826594 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30826847 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30826577 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30826552 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30826700 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12889318 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30826571 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30826620 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30826561 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30826856 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12623891 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30826750 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30826647 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12947626 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30826807 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30826815 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30826763 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30826610 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30826635 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30826822 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30826572 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30826765 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30826566 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30826808 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30826844 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30826650 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30826833 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30826821 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30826764 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30826579 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12834286 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30826559 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30826599 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30826852 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30826702 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30826626 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12622545 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30826643 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30826627 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30826625 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30826601 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30826606 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30826756 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30826680 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30826692 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30826622 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30826858 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12550416 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30826829 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30826597 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30826632 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12942178 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30826602 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30826813 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30826553 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12596209 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30826639 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30826753 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30826590 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30826672 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12501193 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12615599 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30826609 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30826675 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12834290 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30826544 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30826619 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30826843 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30826836 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30826567 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30826659 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30826587 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30826811 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12501192 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30826612 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30826563 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30826825 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30826863 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30826798 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30826758 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12519459 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30826682 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30826555 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30826860 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30826854 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30826576 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30826589 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30826816 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30826604 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30826827 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30826603 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12889325 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30826662 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30826666 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30826665 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30826652 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10436492 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10436491 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1455299_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1453593_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1430596_PM_s_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10436487 | - | - |
| Mouse Genome 430 2.0 Array | 1455299_at | - | - |
| Mouse Genome 430 2.0 Array | 1430596_PM_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1453593_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1430596_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1453593_at | - | - |
| Mouse Genome 430 2.0 Array | 1455299_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 73569_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 73569_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1600006814.mm.1 | NM_028572.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600006814.mm.1 | NM_028572.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600006814.mm.1 | TC1600000797.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1600006815.mm.1 | NM_028572.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600006815.mm.1 | NM_028572.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600006815.mm.1 | TC1600000797.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1600006816.mm.1 | NM_028572.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600006816.mm.1 | NM_028572.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600006816.mm.1 | TC1600000797.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1455299_at | - | - |
| Mouse Expression Set 430 | 1453593_at | - | - |
| Mouse Expression Set 430 | 1430596_PM_s_at | - | - |
| Mouse Expression Set 430 | 1430596_s_at | - | - |
| Mouse Expression Set 430 | 1455299_PM_at | - | - |
| Mouse Expression Set 430 | 1453593_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1453593_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1455299_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1430596_PM_s_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1455299_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1430596_s_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1453593_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 589735 | - | - |

Vgll3
Genomic Map