Assay Details
| Gene Symbol: Bbs9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178415.1 | NP_848502.1 | 12 - 13 | 1683 | 101 | |
| NM_181316.4 | NP_851833.2 | 11 - 12 | 1627 | 101 | ||
| GenBank mRNA | AK028200.1 | - | 11 - 12 | 1717 | 101 | |
| AK086114.1 | - | 11 - 12 | 1599 | 101 | ||
| AK144730.1 | - | 12 - 13 | 1654 | 101 | ||
| AK166297.1 | - | 11 - 12 | 1608 | 101 | ||
| BC030419.1 | - | 8 - 9 | 976 | 101 | ||
| BC046426.1 | - | 11 - 12 | 1639 | 3601505 | 101 | |
More Information
Updated Mapping Information
Set Membership: |
|

Bbs9
Genomic Map