Assay Details
| Gene Symbol: Tmem39a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205286.1 | NP_001192215.1 | 5 - 6 | 716 | 90 | |
| NM_026407.3 | NP_080683.2 | 5 - 6 | 968 | 90 | ||
| GenBank mRNA | AK017817.1 | - | 5 - 6 | 926 | 90 | |
| AK133638.1 | - | 5 - 6 | 940 | 90 | ||
| AK160576.1 | - | 5 - 6 | 717 | 90 | ||
| BC020318.2 | - | 5 - 6 | 923 | 5250532 | 90 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tmem39a
Genomic Map