Assay Details
| Gene Symbol: Tmem39a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205286.1 | NP_001192215.1 | 7 - 8 | 1243 | 66 | |
| NM_001205287.1 | NP_001192216.1 | 7 - 8 | 1291 | 66 | ||
| NM_026407.3 | NP_080683.2 | 7 - 8 | 1495 | 66 | ||
| GenBank mRNA | AK011646.1 | - | 7 - 8 | 1472 | 66 | |
| AK017817.1 | - | 7 - 8 | 1453 | 66 | ||
| AK133638.1 | - | 7 - 8 | 1467 | 66 | ||
| AK160576.1 | - | 7 - 8 | 1244 | 66 | ||
| AK169127.1 | - | 7 - 8 | 1293 | 66 | ||
| BC020318.2 | - | 7 - 8 | 1450 | 5250532 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tmem39a
Genomic Map