Assay Details
| Gene Symbol: Maats1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081025.1 | NP_001074494.1 | 1 - 2 | 202 | 75 | |
| GenBank mRNA | AF503943.1 | - | 1 - 2 | 485 | 75 | |
| AK045121.1 | - | 1 - 2 | 154 | 75 | ||
| AK077038.1 | - | 1 - 2 | 188 | 75 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Maats1
Genomic Map