Assay Details
| Gene Symbol: Maats1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081025.1 | NP_001074494.1 | 4 - 5 | 524 | 114 | |
| GenBank mRNA | AF503943.1 | - | 4 - 5 | 807 | 113 | |
| AK045121.1 | - | 4 - 5 | 476 | 114 | ||
| AK077038.1 | - | 4 - 5 | 510 | 114 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Maats1
Genomic Map