Assay Details
| Gene Symbol: Tmem45a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_183281.2 | NP_899104.2 | 1 - 2 | 62 | 106 | |
| GenBank mRNA | AK009173.1 | - | 1 - 2 | 106 | ||
| AK036849.1 | - | 1 - 2 | 89 | 106 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK009173.1, NM_183281.2 transcripts.
Set Membership: |
|

Tmem45a2
Genomic Map