| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1600014252.mm.1 |
NM_183281.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600014252.mm.1 |
NM_183281.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600014252.mm.1 |
TC1600001755.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1600014255.mm.1 |
TC1600001755.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1600014256.mm.1 |
NM_183281.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600014256.mm.1 |
NM_183281.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600014256.mm.1 |
TC1600001755.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1600014253.mm.1 |
NM_183281.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600014253.mm.1 |
NM_183281.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600014253.mm.1 |
TC1600001755.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1600014254.mm.1 |
NM_183281.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600014254.mm.1 |
NM_183281.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600014254.mm.1 |
TC1600001755.mm.1 |
MTA TC |