Assay Details
| Gene Symbol: Tmem45a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_183281.2 | NP_899104.2 | 5 - 6 | 793 | 83 | |
| GenBank mRNA | AK009173.1 | - | 5 - 6 | 799 | 83 | |
| AK036849.1 | - | 4 - 5 | 2338 | 83 | ||
| BC150914.1 | - | 4 - 5 | 946 | 83 | ||
| BC150924.1 | - | 4 - 5 | 946 | 83 | ||
| BC172084.1 | - | 4 - 5 | 946 | 9054370 | 83 | |
| CN831346.1 | - | 2 - 3 | 206 | 83 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tmem45a2
Genomic Map