Assay Details
| Gene Symbol: Supt5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013676.1 | NP_038704.1 | 11 - 12 | 901 | 60 | |
| GenBank mRNA | AK145117.1 | - | 11 - 12 | 941 | 60 | |
| AK146055.1 | - | 11 - 12 | 929 | 60 | ||
| AK146453.1 | - | 11 - 12 | 929 | 60 | ||
| AK146583.1 | - | 10 - 11 | 1148 | 60 | ||
| AK146650.1 | - | 11 - 12 | 942 | 60 | ||
| AK159786.1 | - | 12 - 13 | 1026 | 60 | ||
| AK178008.1 | - | 1 - 2 | 138 | 60 | ||
| AK187681.1 | - | 1 - 2 | 138 | 60 | ||
| BC007132.1 | - | 11 - 12 | 901 | 3496691 | 60 | |
| BC057449.1 | - | 11 - 12 | 916 | 60 | ||
| BC058598.1 | - | 10 - 11 | 876 | 6306109 | 60 | |
| BC059849.1 | - | 3 - 4 | 484 | 5696323 | 60 | |
| U88539.1 | - | 11 - 12 | 928 | 60 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Supt5
Genomic Map