Assay Details
| Gene Symbol: Supt5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013676.1 | NP_038704.1 | 20 - 21 | 1981 | 72 | |
| GenBank mRNA | AK145117.1 | - | 20 - 21 | 2021 | 72 | |
| AK146055.1 | - | 20 - 21 | 2009 | 72 | ||
| AK146453.1 | - | 20 - 21 | 2009 | 72 | ||
| AK146583.1 | - | 19 - 20 | 2228 | 72 | ||
| AK146650.1 | - | 20 - 21 | 2022 | 72 | ||
| AK159786.1 | - | 21 - 22 | 2106 | 72 | ||
| BC007132.1 | - | 20 - 21 | 1981 | 3496691 | 72 | |
| BC057449.1 | - | 20 - 21 | 1996 | 72 | ||
| BC058598.1 | - | 19 - 20 | 1956 | 6306109 | 72 | |
| BC059849.1 | - | 12 - 13 | 1564 | 5696323 | 72 | |
| U88539.1 | - | 20 - 21 | 2008 | 72 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Supt5
Genomic Map