Assay Details
| Gene Symbol: Supt5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013676.1 | NP_038704.1 | 3 - 4 | 266 | 80 | |
| GenBank mRNA | AK145117.1 | - | 3 - 4 | 306 | 80 | |
| AK145465.1 | - | 3 - 4 | 258 | 80 | ||
| AK146055.1 | - | 3 - 4 | 294 | 80 | ||
| AK146453.1 | - | 3 - 4 | 294 | 80 | ||
| AK146583.1 | - | 2 - 3 | 513 | 80 | ||
| AK146650.1 | - | 3 - 4 | 307 | 80 | ||
| AK159786.1 | - | 4 - 5 | 391 | 80 | ||
| AK194240.1 | - | 1 - 2 | 171 | 80 | ||
| BC007132.1 | - | 3 - 4 | 266 | 3496691 | 80 | |
| BC057449.1 | - | 3 - 4 | 281 | 80 | ||
| BC058598.1 | - | 2 - 3 | 241 | 6306109 | 80 | |
| U88539.1 | - | 3 - 4 | 293 | 80 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Supt5
Genomic Map