Assay Details
| Gene Symbol: Supt5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013676.1 | NP_038704.1 | 10 - 11 | 655 | 91 | |
| GenBank mRNA | AK047062.1 | - | 2 - 3 | 139 | 91 | |
| AK145117.1 | - | 10 - 11 | 695 | 91 | ||
| AK145465.1 | - | 10 - 11 | 647 | 91 | ||
| AK146055.1 | - | 10 - 11 | 683 | 91 | ||
| AK146453.1 | - | 10 - 11 | 683 | 91 | ||
| AK146583.1 | - | 9 - 10 | 902 | 91 | ||
| AK146650.1 | - | 10 - 11 | 696 | 91 | ||
| AK159786.1 | - | 11 - 12 | 780 | 91 | ||
| BC007132.1 | - | 10 - 11 | 655 | 3496691 | 91 | |
| BC057449.1 | - | 10 - 11 | 670 | 91 | ||
| BC058598.1 | - | 9 - 10 | 630 | 6306109 | 91 | |
| BC059849.1 | - | 2 - 3 | 238 | 5696323 | 91 | |
| U88539.1 | - | 10 - 11 | 682 | 91 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Supt5
Genomic Map