Assay Details
| Gene Symbol: Cep19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025892.2 | NP_080168.1 | 1 - 2 | 306 | 78 | |
| GenBank mRNA | AK005324.1 | - | 1 - 2 | 306 | 78 | |
| AK007713.1 | - | 1 - 2 | 290 | 78 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK005324.1, AK007713.1, NM_025892.2 transcripts.
Set Membership: |
|

Cep19
Genomic Map