Assay Details
| Gene Symbol: Pak2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177326.3 | NP_796300.1 | 11 - 12 | 1331 | 85 | |
| GenBank mRNA | AK037275.1 | - | 11 - 12 | 1333 | 85 | |
| AK220528.1 | - | 2 - 3 | 160 | 85 | ||
| AY167030.1 | - | 10 - 11 | 1057 | 85 | ||
| BC043712.1 | - | 1 - 2 | 60 | 85 | ||
| BC063068.1 | - | 1319 | 85 | |||
| BC086650.1 | - | 11 - 12 | 1262 | 6849008 | 85 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pak2
Genomic Map