Assay Details
| Gene Symbol: Pak2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177326.3 | NP_796300.1 | 2 - 3 | 465 | 111 | |
| GenBank mRNA | AK037275.1 | - | 2 - 3 | 466 | 111 | |
| AK190087.1 | - | 1 - 2 | 115 | 111 | ||
| AY167030.1 | - | 1 - 2 | 191 | 111 | ||
| BC063068.1 | - | 453 | 111 | |||
| BC086650.1 | - | 2 - 3 | 396 | 6849008 | 111 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pak2
Genomic Map