Assay Details
| Gene Symbol: Pak2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177326.3 | NP_796300.1 | 9 - 10 | 1101 | 92 | |
| GenBank mRNA | AK037275.1 | - | 9 - 10 | 1103 | 92 | |
| AY167030.1 | - | 8 - 9 | 827 | 92 | ||
| BC063068.1 | - | 1089 | 92 | |||
| BC086650.1 | - | 9 - 10 | 1032 | 6849008 | 92 | |
More Information
Set Membership: |
|

Pak2
Genomic Map