Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1600004005.mm.1 |
NM_026898.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600004005.mm.1 |
NM_026898.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600004005.mm.1 |
TC1600000434.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1600004006.mm.1 |
NM_026898.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600004006.mm.1 |
NM_026898.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600004006.mm.1 |
TC1600000434.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1600004010.mm.1 |
NM_001185162.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600004010.mm.1 |
NM_001185162.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600004010.mm.1 |
NM_026898.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600004010.mm.1 |
NM_026898.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600004010.mm.1 |
TC1600000434.mm.1 |
MTA TC |