Assay Details
| Gene Symbol: Osbpl11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_176840.3 | NP_789810.2 | 2 - 3 | 995 | 98 | |
| GenBank mRNA | BC035278.1 | - | 2 - 3 | 995 | 5043236 | 98 |
| BC099400.1 | - | 2 - 3 | 910 | 98 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Osbpl11
Genomic Map