Assay Details
| Gene Symbol: Osbpl11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_176840.3 | NP_789810.2 | 4 - 5 | 1250 | 85 | |
| GenBank mRNA | BC035278.1 | - | 4 - 5 | 1250 | 5043236 | 85 |
| BC099400.1 | - | 4 - 5 | 1165 | 85 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Osbpl11
Genomic Map