Assay Details
| Gene Symbol: Osbpl11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_176840.3 | NP_789810.2 | 7 - 8 | 1785 | 67 | |
More Information
Updated Mapping Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Osbpl11| Gene Symbol: Osbpl11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_176840.3 | NP_789810.2 | 7 - 8 | 1785 | 67 | |
Set Membership: |
|