Assay Details
| Gene Symbol: Map2k5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011840.2 | NP_035970.1 | 12 - 13 | 1393 | 62 | |
| GenBank mRNA | AB019374.1 | - | 12 - 13 | 1394 | 62 | |
| AK013856.1 | - | 4 - 5 | 236 | 62 | ||
| AK159261.1 | - | 12 - 13 | 1443 | 62 | ||
| BC028260.1 | - | 12 - 13 | 1273 | 5067818 | 62 | |
More Information
Updated Mapping Information
Set Membership: |
|

Map2k5
Genomic Map