Assay Details
| Gene Symbol: Map2k5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011840.2 | NP_035970.1 | 19 - 20 | 1731 | 57 | |
| GenBank mRNA | AB019374.1 | - | 19 - 20 | 1732 | 57 | |
| AK159261.1 | - | 19 - 20 | 1781 | 57 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Map2k5
Genomic Map