| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0900020776.mm.1 |
NM_016769.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900020776.mm.1 |
NM_016769.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900020776.mm.1 |
TC0900002480.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900020784.mm.1 |
NM_016769.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900020784.mm.1 |
NM_016769.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900020784.mm.1 |
TC0900002480.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900020777.mm.1 |
NM_016769.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900020777.mm.1 |
NM_016769.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900020777.mm.1 |
TC0900002480.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900020783.mm.1 |
NM_016769.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900020783.mm.1 |
NM_016769.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900020783.mm.1 |
TC0900002480.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900020785.mm.1 |
NM_016769.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900020785.mm.1 |
NM_016769.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900020785.mm.1 |
TC0900002480.mm.1 |
MTA TC |