Assay Details
| Gene Symbol: Iqch | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030068.1 | NP_084344.1 | 19 - 20 | 2856 | 93 | |
| GenBank mRNA | AK019535.1 | - | 19 - 20 | 2975 | 93 | |
| BC141047.1 | - | 19 - 20 | 2856 | 9055659 | 93 | |
More Information
Updated Mapping Information
Set Membership: |
|

Iqch
Genomic Map