Assay Details
| Gene Symbol: Iqch | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030068.1 | NP_084344.1 | 5 - 6 | 521 | 74 | |
| GenBank mRNA | AK019535.1 | - | 5 - 6 | 553 | 74 | |
| AK029502.1 | - | 5 - 6 | 557 | 74 | ||
| BC141047.1 | - | 5 - 6 | 521 | 9055659 | 74 | |
| BC145231.1 | - | 5 - 6 | 521 | 9053783 | 74 | |
More Information
Updated Mapping Information
Set Membership: |
|

Iqch
Genomic Map