Assay Details
| Gene Symbol: Mapk6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_015806.5 | NP_056621.4 | 2 - 3 | 1329 | 84 | |
| NM_027418.2 | NP_081694.1 | 2 - 3 | 1333 | 84 | ||
| GenBank mRNA | AF132850.1 | - | 1 - 2 | 1062 | 84 | |
| AK044752.1 | - | 2 - 3 | 1311 | 84 | ||
| BC006778.1 | - | 1 - 2 | 543 | 84 | ||
| BC037060.1 | - | 1 - 2 | 1067 | 84 | ||
| BC048779.1 | - | 1 - 2 | 831 | 6315375 | 84 | |
| BC052420.1 | - | 1 - 2 | 578 | 5702905 | 84 | |
| BC100385.1 | - | 2 - 3 | 1311 | 30732053 | 84 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mapk6
Genomic Map