Assay Details
| Gene Symbol: Mapk6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_015806.5 | NP_056621.4 | 4 - 5 | 1638 | 75 | |
| NM_027418.2 | NP_081694.1 | 4 - 5 | 1642 | 75 | ||
| GenBank mRNA | AF132850.1 | - | 3 - 4 | 1371 | 75 | |
| BC006778.1 | - | 3 - 4 | 852 | 75 | ||
| BC037060.1 | - | 3 - 4 | 1376 | 75 | ||
| BC048779.1 | - | 3 - 4 | 1140 | 6315375 | 75 | |
| BC052420.1 | - | 3 - 4 | 887 | 5702905 | 75 | |
| BC100385.1 | - | 4 - 5 | 1620 | 30732053 | 75 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mapk6
Genomic Map