Assay Details
| Gene Symbol: Bco2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133217.3 | NP_573480.1 | 8 - 9 | 1208 | 106 | |
| GenBank mRNA | AJ290392.1 | - | 7 - 8 | 1051 | 106 | |
| BC107006.2 | - | 7 - 8 | 1051 | 40046913 | 106 | |
| BC107007.2 | - | 7 - 8 | 1051 | 40046914 | 106 | |
More Information
Updated Mapping Information
Set Membership: |
|

Bco2
Genomic Map