Assay Details
| Gene Symbol: Lztr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025808.3 | NP_080084.2 | 2 - 3 | 306 | 88 | |
| GenBank mRNA | AK004561.2 | - | 2 - 3 | 306 | 88 | |
| AK005037.1 | - | 2 - 3 | 305 | 88 | ||
| AK083411.1 | - | 2 - 3 | 310 | 88 | ||
| AK132817.1 | - | 2 - 3 | 305 | 88 | ||
More Information
Set Membership: |
|

Lztr1
Genomic Map