Assay Details
| Gene Symbol: Lztr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025808.3 | NP_080084.2 | 6 - 7 | 640 | 64 | |
| GenBank mRNA | AK004561.2 | - | 6 - 7 | 640 | 64 | |
| AK005037.1 | - | 6 - 7 | 639 | 64 | ||
| AK083411.1 | - | 6 - 7 | 644 | 64 | ||
| AK132817.1 | - | 6 - 7 | 639 | 64 | ||
| AK215859.1 | - | 2 - 3 | 200 | 64 | ||
| BC034400.1 | - | 6 - 7 | 637 | 2651456 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|

Lztr1
Genomic Map