Assay Details
| Gene Symbol: Spag6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_015773.2 | NP_056588.1 | 4 - 5 | 616 | 57 | |
| GenBank mRNA | AF173866.1 | - | 4 - 5 | 553 | 57 | |
| AK005732.1 | - | 4 - 5 | 564 | 57 | ||
| AK054199.1 | - | 3 - 4 | 652 | 57 | ||
| BC090660.1 | - | 4 - 5 | 559 | 30673350 | 57 | |
| BC100455.1 | - | 4 - 5 | 566 | 30919921 | 57 | |
More Information
Updated Mapping Information
Set Membership: |
|

Spag6
Genomic Map