Assay Details
| Gene Symbol: Spag6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_015773.2 | NP_056588.1 | 6 - 7 | 997 | 111 | |
| GenBank mRNA | AF173866.1 | - | 6 - 7 | 934 | 111 | |
| AK005732.1 | - | 6 - 7 | 945 | 111 | ||
| AK054199.1 | - | 5 - 6 | 1033 | 111 | ||
| BC090660.1 | - | 6 - 7 | 940 | 30673350 | 111 | |
| BC100455.1 | - | 6 - 7 | 947 | 30919921 | 111 | |
More Information
Set Membership: |
|

Spag6
Genomic Map