Assay Details
| Gene Symbol: Mapk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038663.1 | NP_001033752.1 | 2 - 3 | 530 | 64 | |
| NM_011949.3 | NP_036079.1 | 2 - 3 | 530 | 64 | ||
| GenBank mRNA | AK035386.1 | - | 2 - 3 | 504 | 64 | |
| AK048127.1 | - | 2 - 3 | 532 | 64 | ||
| AK087925.1 | - | 2 - 3 | 505 | 64 | ||
| AK132241.1 | - | 2 - 3 | 518 | 64 | ||
| AK148853.1 | - | 2 - 3 | 510 | 64 | ||
| BB648734.1 | - | 2 - 3 | 531 | 64 | ||
| BC058258.1 | - | 2 - 3 | 473 | 6468233 | 64 | |
| D10939.1 | - | 2 - 3 | 345 | 64 | ||
| X58712.1 | - | 2 - 3 | 303 | 64 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mapk1
Genomic Map