Assay Details
| Gene Symbol: Mapk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038663.1 | NP_001033752.1 | 3 - 4 | 732 | 82 | |
| NM_011949.3 | NP_036079.1 | 3 - 4 | 732 | 82 | ||
| GenBank mRNA | AK035386.1 | - | 3 - 4 | 706 | 82 | |
| AK048127.1 | - | 3 - 4 | 734 | 82 | ||
| AK087925.1 | - | 3 - 4 | 707 | 82 | ||
| AK132241.1 | - | 3 - 4 | 720 | 82 | ||
| AK148853.1 | - | 3 - 4 | 712 | 82 | ||
| AK193161.1 | - | 1 - 2 | 62 | 82 | ||
| BC058258.1 | - | 3 - 4 | 675 | 6468233 | 82 | |
| D10939.1 | - | 3 - 4 | 547 | 82 | ||
| X58712.1 | - | 3 - 4 | 505 | 82 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mapk1
Genomic Map