Assay Details
| Gene Symbol: Aifm3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291070.1 | NP_001277999.1 | 3 - 4 | 505 | 105 | |
| NM_175178.4 | NP_780387.2 | 5 - 6 | 911 | 105 | ||
| GenBank mRNA | AK012958.1 | - | 2 - 3 | 172 | 105 | |
| AK158809.1 | - | 4 - 5 | 573 | 105 | ||
| BC046517.1 | - | 5 - 6 | 897 | 105 | ||
| BC096476.1 | - | 4 - 5 | 686 | 4221224 | 105 | |
| BC120685.1 | - | 4 - 5 | 540 | 40129608 | 105 | |
More Information
Updated Mapping Information
Set Membership: |
|

Aifm3
Genomic Map