Assay Details
| Gene Symbol: Aifm3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291070.1 | NP_001277999.1 | 7 - 8 | 867 | 66 | |
| NM_175178.4 | NP_780387.2 | 9 - 10 | 1273 | 66 | ||
| GenBank mRNA | AK012958.1 | - | 5 - 6 | 625 | 66 | |
| AK049928.1 | - | 9 - 10 | 1104 | 66 | ||
| AK158809.1 | - | 8 - 9 | 935 | 66 | ||
| BC046517.1 | - | 9 - 10 | 1259 | 66 | ||
| BC096476.1 | - | 8 - 9 | 1048 | 4221224 | 66 | |
| BC120685.1 | - | 8 - 9 | 902 | 40129608 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|

Aifm3
Genomic Map