Assay Details
| Gene Symbol: Arih1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019927.2 | NP_064311.2 | 10 - 11 | 1180 | 65 | |
| GenBank mRNA | AJ130977.1 | - | 10 - 11 | 877 | 65 | |
| BC038034.1 | - | 10 - 11 | 1100 | 4035861 | 65 | |
| BC057680.1 | - | 10 - 11 | 1420 | 3498094 | 65 | |
| CK633971.1 | - | 1 - 2 | 75 | 65 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Arih1
Genomic Map