Assay Details
| Gene Symbol: Arih1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019927.2 | NP_064311.2 | 12 - 13 | 1500 | 106 | |
| GenBank mRNA | AJ130977.1 | - | 12 - 13 | 1200 | 109 | |
| AK087802.1 | - | 2 - 3 | 332 | 106 | ||
| BC038034.1 | - | 12 - 13 | 1420 | 4035861 | 106 | |
| BC057680.1 | - | 12 - 13 | 1740 | 3498094 | 106 | |
| CK633971.1 | - | 3 - 4 | 395 | 106 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Arih1
Genomic Map