Assay Details
| Gene Symbol: Arih1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019927.2 | NP_064311.2 | 6 - 7 | 831 | 123 | |
| GenBank mRNA | AF361001.1 | - | 6 - 7 | 545 | 123 | |
| AJ130977.1 | - | 6 - 7 | 528 | 123 | ||
| BC038034.1 | - | 6 - 7 | 751 | 4035861 | 123 | |
| BC057680.1 | - | 6 - 7 | 1071 | 3498094 | 123 | |
| CV555932.1 | - | 6 - 7 | 486 | 123 | ||
More Information
Set Membership: |
|

Arih1
Genomic Map