Assay Details
| Gene Symbol: Arih1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019927.2 | NP_064311.2 | 8 - 9 | 975 | 108 | |
| GenBank mRNA | AJ130977.1 | - | 8 - 9 | 672 | 108 | |
| BC038034.1 | - | 8 - 9 | 895 | 4035861 | 108 | |
| BC057680.1 | - | 8 - 9 | 1215 | 3498094 | 108 | |
More Information
Updated Mapping Information
Set Membership: |
|

Arih1
Genomic Map