Assay Details
| Gene Symbol: Parp6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205239.1 | NP_001192168.1 | 11 - 12 | 1365 | 64 | |
| NM_029922.3 | NP_084198.2 | 11 - 12 | 1305 | 64 | ||
| NR_038074.1 | 11 - 12 | 1305 | 64 | |||
| NR_038075.1 | 11 - 12 | 1365 | 64 | |||
| GenBank mRNA | AK035349.1 | - | 11 - 12 | 1229 | 64 | |
| AK075876.1 | - | 1 - 2 | 67 | 64 | ||
| AK076321.1 | - | 11 - 12 | 1229 | 64 | ||
| BC031387.1 | - | 10 - 11 | 1036 | 5038718 | 64 | |
| BC052064.1 | - | 11 - 12 | 1303 | 64 | ||
| BC058266.1 | - | 2 - 3 | 259 | 64 | ||
| BC062096.1 | - | 10 - 11 | 1222 | 5716648 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|

Parp6
Genomic Map