Assay Details
| Gene Symbol: Parp6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205239.1 | NP_001192168.1 | 16 - 17 | 1819 | 59 | |
| NM_029922.3 | NP_084198.2 | 16 - 17 | 1759 | 59 | ||
| NR_038074.1 | 16 - 17 | 1759 | 59 | |||
| NR_038075.1 | 16 - 17 | 1819 | 59 | |||
| GenBank mRNA | AK009530.1 | - | 6 - 7 | 519 | 59 | |
| AK014142.1 | - | 7 - 8 | 562 | 59 | ||
| AK035349.1 | - | 16 - 17 | 1683 | 59 | ||
| AK075876.1 | - | 6 - 7 | 521 | 59 | ||
| AK076321.1 | - | 16 - 17 | 1683 | 59 | ||
| BC030184.1 | - | 2 - 3 | 109 | 59 | ||
| BC031387.1 | - | 15 - 16 | 1490 | 5038718 | 59 | |
| BC052064.1 | - | 16 - 17 | 1757 | 59 | ||
| BC058266.1 | - | 7 - 8 | 713 | 59 | ||
| BC062096.1 | - | 15 - 16 | 1676 | 5716648 | 59 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC030184.1 transcripts.
Set Membership: |
|

Parp6
Genomic Map