Assay Details
| Gene Symbol: Slc25a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153150.2 | NP_694790.1 | 2 - 3 | 345 | 79 | |
| GenBank mRNA | AK005070.1 | - | 2 - 3 | 275 | 79 | |
| AK159289.1 | - | 2 - 3 | 308 | 79 | ||
| BC037087.1 | - | 2 - 3 | 305 | 5124403 | 79 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Slc25a1
Genomic Map