Assay Details
| Gene Symbol: Slc25a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153150.2 | NP_694790.1 | 6 - 7 | 777 | 65 | |
| GenBank mRNA | AK005070.1 | - | 6 - 7 | 707 | 65 | |
| AK159289.1 | - | 6 - 7 | 740 | 65 | ||
| AK170191.1 | - | 6 - 7 | 741 | 65 | ||
| AK188281.1 | - | 3 - 4 | 245 | 65 | ||
| AK211700.1 | - | 3 - 4 | 245 | 65 | ||
| AK219123.1 | - | 3 - 4 | 244 | 65 | ||
| BC037087.1 | - | 6 - 7 | 737 | 5124403 | 65 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Slc25a1
Genomic Map