Assay Details
| Gene Symbol: Pld3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011116.1 | NP_035246.1 | 11 - 12 | 1429 | 63 | |
| GenBank mRNA | AF026124.1 | - | 11 - 12 | 1429 | 63 | |
| AK153508.1 | - | 9 - 10 | 1118 | 63 | ||
| AK188215.1 | - | 1 - 2 | 80 | 63 | ||
| BC068170.1 | - | 11 - 12 | 1425 | 63 | ||
| BC076586.1 | - | 11 - 12 | 1493 | 30604186 | 63 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25017834 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25017836 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2037513 | BC068170.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2037513 | BC076586.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2037513 | AF026124.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2037513 | AK003045.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2037513 | AK153508.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 521635 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 93029 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 484860 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 539966 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 57352 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 469240 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 365731 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 326314 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 122671 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 197104 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 746704 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 573423 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 865333 | AK017913.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 865333 | XM_985015.1 | refseq_mrna_mouse_XM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 429923 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 834498 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 202681 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 143969 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P142300 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P145152 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2037513 | BC068170.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2037513 | BC076586.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2037513 | AF026124.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2037513 | AK003045.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2037513 | AK153508.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25017835 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25017825 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25017829 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25017822 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25017831 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25017833 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25017819 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25017827 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25017836 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P145152 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P142300 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06147451 | - | - |
| Whole Mouse Genome G4122A | A_52_P614336 | BC068170.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P614336 | BC076586.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P614336 | AF026124.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P614336 | AK003045.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P614336 | AK153508.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P119885 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P142300 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P145152 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06155686 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06155689 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25017834 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25017836 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25017734 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25017792 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25017694 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25017690 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25017742 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25017786 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25017750 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25017700 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25017727 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25017777 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25017714 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25017704 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25017708 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25017825 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25017709 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25017833 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25017813 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25017789 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25017719 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25017732 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25017725 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25017829 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25017805 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25017682 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25017736 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25017811 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25017835 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25017767 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25017769 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25017757 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25017703 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25017770 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25017686 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25017800 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25017685 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25017795 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25017776 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25017764 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25017761 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25017710 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25017797 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25017680 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25017772 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25017806 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25017831 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25017785 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25017760 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25017791 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25017683 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25017808 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25017802 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25017794 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25017738 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25017754 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25017783 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25017684 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25017804 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25017711 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25017779 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25017729 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25017723 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25017698 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25017759 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25017692 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25017746 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25017766 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25017716 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25017688 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25017793 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25017807 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25017781 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25017810 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25017730 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25017713 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25017702 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25017816 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25017822 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25017720 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25017826 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25017774 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25017744 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25017760 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25017732 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06504556 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25017825 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06098866 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25017750 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25017831 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06052288 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25017819 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25017822 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25017738 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06140151 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25017793 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25017734 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25017818 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06381473 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25017748 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25017833 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06109059 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25017806 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25017805 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25017757 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25017797 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25017742 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25017800 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25017795 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06121042 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25017815 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25017736 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06140161 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25017827 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06098864 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25017794 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25017791 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25017816 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25017744 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25017786 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25017758 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25017789 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25017829 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25017802 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25017828 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06075188 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10561319 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10561310 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10561316 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10561322 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10561314 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10561308 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10561313 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10561315 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10561320 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10561321 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10561309 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10561317 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10561318 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10561307 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10561312 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10561311 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1416013_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10561306 | - | - |
| Mouse Genome 430 2.0 Array | 1416013_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1416013_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 18807_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 18807_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0700023297.mm.1 | TC0700002669.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700023297.mm.1 | NM_011116.2 | Refseq |
| Mouse Expression Set 430 | 1416013_PM_at | - | - |
| Mouse Expression Set 430 | 1416013_at | - | - |
| Murine Genome U74v2 Set | 100607_PM_at | - | - |
| Murine Genome U74v2 Set | 100607_at | - | - |
| Mouse Genome 430A 2.0 Array | 1416013_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1416013_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1416013_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1416013_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 105700093 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 105700093 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 510041 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0000510041 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2702101 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2702101 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2543265 | - | - |
| mouse-6 v1.1 (Illumina) | scl31600.13.1_22-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 105700093 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 510041 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl31600.13.1_22-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 105700093 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 510041 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2784950 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2702101 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2784950 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2702101 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2543265 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl31600.13.1_22-S | - | - |

Pld3
Genomic Map